missing translation for 'onlineSavingsMsg'
Learn More

Scopolamine hydrobromide trihydrate, 99%

Catalog No. p-191672
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161750050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161750050 Supplier Thermo Scientific Chemicals Supplier No. 161750050
Only null left

Scopolamine hydrobromide trihydrate, CAS # 6533-68-2, is the trihydrate form of scopolamine hydrobromide that shows competitive anti-muscarinic activity by blocking the actions of endogenous acetylcholine or exogenous agonists. It also shows anti-spasmodic and anti-emetic activities. | CAS: 6533-68-2 | C17H28BrNO7 | 438.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Scopolamine hydrobromide trihydrate is a salt form of scopolamine, a tropane alkaloid derived from plants of the nightshade family (Solanaceae), mainly Hyoscyamus niger and Atropa belladonna
• Scopolamine hydrobromide trihydrate, demonstrates competitive anti-muscarinic activity blocking the actions of endogenous acetylcholine or exogenous agonists
• It also shows anti-spasmodic and anti-emetic activities

Applications

• It has been described that scopolamine inhibits muscarinic acetylcholine receptors (mAChRs) with some selectivity for M1 mAChRs
• It is structurally similar to acetylcholine and can antagonize acetylcholine activity mediated by muscarinic receptors located on structures innervated by postganglionic cholinergic nerves and on smooth muscles (that respond to acetylcholine but lack cholinergic innervation)
• Scopolamine hydrobromide trihydrate can induce amnesia in laboratory animals and so it is frequently used in animal models of memory deficit
• Scopolamine hydrobromide trihydrate is used to induce cognitive deficits in animal models of Alzheimer’s disease
• Scopolamine, has been employed in several studies to understand, identify, and characterize biological targets for Alzheimer's disease

Chemical Identifiers

CAS 6533-68-2
Molecular Formula C17H28BrNO7
Molecular Weight (g/mol) 438.32
MDL Number MFCD00150066
InChI Key LACQPOBCQQPVIT-FCFGRTKISA-N
Synonym hyoscine hydrobromide trihydrate
PubChem CID 131632304
IUPAC Name hydrogen (1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate trihydrate bromide
SMILES [H+].O.O.O.[Br-].CN1[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1

Specifications

Melting Point 194.0°C to 197.0°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 102
Merck Index 14, 8406
Specific Rotation Condition −27° (20°C c=5,H2O)
Specific Rotation − 27.00
Solubility Information Solubility in water: 700g/L water (20 c). Other solubilities: 1g/20 mL alcohol, slightly soluble in chloroform, practically insoluble in ether
Formula Weight 438.3
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Scopolamine hydrobromide trihydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber YM4730000

RUO – Research Use Only