missing translation for 'onlineSavingsMsg'
Learn More

Sclareol 96.0+%, TCI America™

Catalog No. S09161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09161G Supplier TCI America Supplier No. S09161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 515-03-7
Molecular Formula C20H36O2
Molecular Weight (g/mol) 308.506
MDL Number MFCD00869558
InChI Key XVULBTBTFGYVRC-SVPXJYONSA-N
Synonym (1R,2R,8aS)-Decahydro-1-(3-hydroxy-3-methyl-4-pentenyl)-2,5,5,8a-tetramethyl-2-naphthol
PubChem CID 56842011
IUPAC Name (1R,2R,4aS,8aS)-1-(3-hydroxy-3-methylpent-4-enyl)-2,5,5,8a-tetramethyl-3,4,4a,6,7,8-hexahydro-1H-naphthalen-2-ol
SMILES CC1(CCCC2(C1CCC(C2CCC(C)(C=C)O)(C)O)C)C

Specifications

Melting Point 101°C
Color White-Yellow
Boiling Point 189°C
Quantity 1 g
Formula Weight 308.51
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Sclareol
RTECSNumber QK0301900
TSCA Yes