Learn More
World Precision Operating Scissors, 16cm, Sharp/Sharp, Curved, German
Catalog No. 50822569
Encompass
Ring-handled Operating Scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 16 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors are available with straight or curved blades. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved.
Catalog No. 50822569 Supplier World Precision Supplier No. 501228G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ring-handled Operating Scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 16 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors are available with straight or curved blades. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. They are available in both surgical grade stainless steel and our premium German steel, making them more durable and resistant to corrosion. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved.