Learn More
Fine Science Tools SCISSORS LEXOR 10CC
Catalog No. NC9147994
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 26mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC9147994 Supplier Fine Science Tools Supplier No. 1407810
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 26mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No