missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ScintiVerse™ BD Cocktail (Scintanalyzed™), Fisher Chemical
Universal LSC Cocktail for aqueous, nonaqueous and emulsion counting systems. Contains biodegradable linear alkyl aromatic solvent, which can be classified as nonhazardous liquid by local sewerage regulations
Supplier: Fisher Chemical FLSX184
Description
- Universal LSC Cocktail for aqueous, nonaqueous and emulsion counting systems.
- Contains biodegradable linear alkyl aromatic solvent, which can be classified as a nonhazardous liquid by local sewerage regulations.
Specifications
| ScintiVerse™ BD Cocktail | |
| 67774-74-7,577-11-7,9016-45-9,92-71-7,13280-61-0 | |
| 0.9g/mL | |
| 279°F min. | |
| MFCD00212667 | |
| Glass Bottle | |
| 232.41 | |
| 0.890 to 0.910 (at 25°C) |
| Liquid | |
| 50 CPM max. | |
| 4 L | |
| C17H28 | |
| 0.55 min., 0.56 min. | |
| XBEADGFTLHRJRB-UHFFFAOYSA-N | |
| Scintanalyzed | |
| 0.7% Max. |