Learn More
Eppendorf™ Scientific Shaker Universal Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
$1,999.44 - $2,881.14
Specifications
Product Type Universal Platform
Includes Pre-drilled holes
View More Specs
Requires: Clamps (sold separately)
Products 2
Catalog Number Mfr. No. Length (English) Platform Width (English) Platform For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Platform Width (English) Platform For Use With (Equipment) Price Quantity & Availability  
1427842
Accessories View Documents Promotion Details
Eppendorf™
M12509902
18 in. 18 in. Innova, I-Series, Excella Shaker
Each for $1,999.44
Only null left
1427841
Accessories View Documents Promotion Details
Eppendorf™
M12509920
30 in. 18 in. Innova 43/43R, 2300, Series 4300 and 4400, E10, E25, E25R, C10, C25/C25KC Shaker
Each for $2,881.14
Only null left
Description
Description
  • Accept a variety of accessory clamps, test tube racks and gel trays for greater flexibility
  • Hold combinations of flasks or test tube racks
  • With pre-drilled holes
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Specifications
Universal Platform
Pre-drilled holes