Learn More
Scientific Device Laboratory Pinworm Collectors DFS Item
Specially designed cap with paddle for pin worm collection
Supplier:  Scientific Device 582
Catalog No. 23-769-507
Only null left
Explore available promotions
Description
Description
  • Clearer and thinner plastic paddle with sticky side for pinworm collection
  • Paddle can be viewed directly under microscope by snapping off top
Specifications
Specifications
Clear
Cap with Paddle
Pinworm Collector
50/Pk.
Product Suggestions
Product Suggestions