missing translation for 'onlineSavingsMsg'
Learn More

Scientific Device Humidity/Slide Moisture Chamber

Catalog No. 23769522
Encompass
Change view
Click to view available options
Slide moisture chamber, Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23769522 Slide moisture chamber, Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23769522 Supplier Scientific Device Supplier No. 197BL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for both immunostaining and diagnostic reactions performed on slides

  • All units divided into 10 compartments that separated by lid barrier
  • Chambers are made of heavy-duty polystyrene
  • Air tight sealing keep moisture inside
  • Stackable boxes enables easy storage
  • Black in color

Specifications

Type Slide Moisture Chamber
Color Black
Material Heavy Duty Polystyrene
For Use With (Application) Both immunostaining and diagnostic reactions performed on slides
Includes Lid barrier between each slide