Learn More
Scientific Device Acid Fast Control Slides
Catalog No. 23769301
Encompass
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
23769301 50 Pk.
1 options
Catalog No. 23769301 Supplier Scientific Device Supplier No. 351
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Convenient control slides
  • Simulate a clinical sputum smear with appropriate background material
  • Each slide features two wells containing a positive and negative smear
  • May be used with Kinyoun, Ziehl-Neelsen or other fluorescent techniques
Specifications
Quantity 50 Pk.