missing translation for 'onlineSavingsMsg'
Learn More

Supertek™ Science Cube Kit - E-Cube 4 in 1 Kit
GREENER_CHOICE

Catalog No. S89515
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89515 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S89515 Supplier Supertek™ Supplier No. PH96016S3

The E-Cube 4 in 1 Kit has everything from our 4 most popular kits - Telegraph Kit, Color Theory, Simple Circuit, and Conduction Experiment. Explore many aspects of electricity and take it to the next step with our digital multi-meter. Have fun recording your results and design your own experiments.

  • Perfect for Group Experiments
  • Perfect for Homeschooling
  • Save space when storing in reusable plastic snap box
  • Includes:
    • 1 Reusable plastic snap box
    • Five D-cell battery holders
    • 1 knife switch
    • 4 light bulb holders
    • 5 light bulbs
    • 17 alligator cords
    • 2 contact key
    • 1 buzzer
    • 1 miniature motor
    • 8 disks
    • 1 conduction sample pack
    • 1 digital multi-meter
    • 4 Exploration Guide
  • Ages 7+

Specifications

Grade 1
Age 7
For Use With (Application) Designing Experiments and Recording Results
Class Size 1 Lab Group
Product Type Supertek™ Science Cube Kits