missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich Schiff′s Reagent Intense

Catalog No. 01288063
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288063 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288063 Supplier Sigma-Aldrich Supplier No. 1025721007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the detection of aldehyde and mucosubstances in microscopy.

The Schiff's reagent Intense, for the detection of aldehyde and mucosubstances in microscopy, is a ready-to-use solution used for human-medical cell diagnosis and serves the histological investigation of sample material of human origin, for example histological sections of e. g. the intestine or liver.

  • This ready-to-use Schiff's reagent Intense can be used to stain mucopolysaccharides in histological tissue specimens. The PAS (periodic acid schiff) reaction is one of the most frequently used chemical methods for the detection of aldehyde and mucosubstances in microscopy.
  • In the PAS reaction, the histological specimen material is first treated with periodic acid, resulting in the oxidation of the 1,2-glycols into aldehyde groups.
  • The addition of Schiff's reagent (fuchsin-sulfuric acid) in the second step causes the aldehydes to react to form a brilliant red color.
  • In the end result, the PAS reaction yields a specific color reaction with unsubstituted polysaccharides, neutral mucopolysaccharides, muco- and glycoproteins, and glyco- and phospholipids.
  • The PAS reaction can be further combined with the Alcian blue staining method to detect mucosubstances (glycosaminoglycans).
  • The 500 mL bottle is sufficient for 3000 stainings.
  • Form: liquid
  • IVD: for in vitro diagnostic use
  • pH: 1.74 in H2O
  • Storage temperature: 20°C to 25°C.

Specifications

Quantity 500 mL
Product Type Schiff′s Reagent Intense