missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Catalog No. AA40579NA
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40579NA 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40579NA Supplier Thermo Scientific Chemicals Supplier No. 40579NA
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 14066-05-8
Molecular Formula Cl3O12Sc
Molecular Weight (g/mol) 343.294
MDL Number MFCD00243289
InChI Key UBVKRMCXDCYZQK-UHFFFAOYSA-K
Synonym scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorate
PubChem CID 11013318
IUPAC Name scandium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3]

Specifications

Physical Form Liquid
Quantity 2 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 451.42 (343.30 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Scandium(III) perchlorate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-914-6
TSCA No
Recommended Storage Ambient temperatures