missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) nitrate hydrate, REacton™, 99.99% (REO)

Catalog No. AA1122103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1122103 1 g
AA1122106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1122103 Supplier Thermo Scientific Chemicals Supplier No. 01122103
Only null left

CAS: 107552-14-7 | N3O9Sc | 230.968 g/mol

Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107552-14-7
Molecular Formula N3O9Sc
Molecular Weight (g/mol) 230.968
MDL Number MFCD00011220
InChI Key DFCYEXJMCFQPPA-UHFFFAOYSA-N
Synonym scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt
PubChem CID 166818
IUPAC Name scandium(3+);trinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sc+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 1 g
UN Number UN1477
Merck Index 14,8392
Solubility Information Soluble in water and alcohol.
Formula Weight 230.97 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 236-701-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only