Learn More
Scandium(III) nitrate hydrate, REacton™, 99.9% (REO)
CAS: 107552-14-7 | N3O9Sc | 230.97 g/mol
$236.55 - $824.40
Chemical Identifiers
| CAS | 107552-14-7 |
|---|---|
| Molecular Formula | N3O9Sc |
| MDL Number | MFCD00011220 |
| InChI Key | DFCYEXJMCFQPPA-UHFFFAOYSA-N |
| Synonym | scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt |
| PubChem CID | 166818 |
| IUPAC Name | scandium(3+);trinitrate |
Description
Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 107552-14-7 | |
| MFCD00011220 | |
| scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt | |
| scandium(3+);trinitrate |
| N3O9Sc | |
| DFCYEXJMCFQPPA-UHFFFAOYSA-N | |
| 166818 |
Specifications
| 107552-14-7 | |
| Crystalline | |
| N3O9Sc | |
| UN1477 | |
| scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt | |
| DFCYEXJMCFQPPA-UHFFFAOYSA-N | |
| scandium(3+);trinitrate | |
| 166818 | |
| 99.9% | |
| Hygroscopic | |
| (REO) |
| White | |
| 1 g | |
| MFCD00011220 | |
| 14,8392 | |
| Soluble in water and alcohol. | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Sc+3] | |
| 230.968 | |
| 230.97 (Anhydrous) | |
| Reacton | |
| Scandium (III) nitrate hydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber : 236-701-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only