missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) Acetate Hydrate 98.0+%, TCI America™

Catalog No. S05341G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05341G Supplier TCI America Supplier No. S05341G
Only null left

Specifications

CAS 207671-51-0
Color White
Molecular Formula C6H9O6Sc
Quantity 1 g
MDL Number MFCD00150122
Synonym Acetic Acid Scandium(III) Salt
Formula Weight 222.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Scandium(III) Acetate Hydrate