missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience Scan 500
Description
Scan 500 is an automatic colony counter that ensures fast and efficient counting of colonies, including on chromogenic agar. It guarantees excellent reproducibility. The images and results are saved and usable anytime. Scan 500 is also an inhibition zone reader.
- Minimum size of detected colony: 0.1 mm
- Camera: 1 megapixel to zoom x 28
- Counting on 55 to 90 mm dia. round Petri dishes
- Counting on pour, surface, Spiral, circle mode plated Petri dishes
- Counting on chromogenic agar
- Inhibition zone reader
Specifications
Specifications
| Product Type | Automated Colony Counter and Zone Measurement System |
| Display Type | LED |
| Voltage | 100/240 V |