missing translation for 'onlineSavingsMsg'
Learn More

Interscience Scan 4000

The Scan 4000 is an ultra-HD automatic colony counter and inhibition zone reader with unparalleled image quality.

Supplier:  Interscience 438000

Catalog No. 16-949-091

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Scan 4000 is an automatic HD colour colony counter. It is adapted to all sizes of Petri dishes and any culture media. Its indirect lighting system gives great user comfort, high accuracy and excellent reproducibility. Scan 4000 is also an inhibition zone reader.

  • Minimum size of detected colony: 0.05 mm
  • Camera: 5 megapixels to zoom x 69
  • Counting on 55 to 90 dia. mm round Petri dishes and 120 mm square Petri dishes
  • Counting on pour, surface, Spiral, circle mode plated dishes
  • Counting on chromogenic agar, Petrifilm™, MC-media Pads™, Compact Dry™, easyPlate™, filtration membranes
  • Inhibition zone reader
Specifications

Specifications

Automated Colony Counter and Zone Measurement System
LED
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions