missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience Scan 1200
Description
Scan 1200 is an HD automatic colony counter. It adapts itself to all media and counts in 1 click all colonies even the smallest ones. It ensures accuracy and excellent reproducibility. Images and results are saved automatically. Scan 1200 is also an inhibition zone reader.
- Minimum size of detected colony: 0.05 mm
- Camera: 1.2 megapixels to zoom x 28
- Counting on 55 to 90 mm dia. round Petri dishes
- Counting on pour, surface, Spiral, circle mode plated Petri dishes
- Counting on chromogenic agar, Petrifilm™, MC-media Pads™, Compact Dry™, EasyPlate™, filtration membranes
- Inhibition zone reader
Specifications
Specifications
| Product Type | Automated Colony Counter and Zone Measurement System |
| Display Type | LED |
| Voltage | 100/240 V |