Learn More
Mopec Scalpel Blades, Double-Edged, 100 pack 
Catalog No. AJ111
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
AJ111 10/Pk.
1 options
Catalog No. AJ111 Supplier Mopec Supplier No. AJ111
Mopec's Double-edged blade is ideal for trimming surgical specimens. A versatile tool for any laboratory.
  • Double-Edged
  • Versatile
  • Ideal for trimming surgical specimens.
Specifications
Blade Material Stainless Steel
Product Type Scalpel Blade
Size 2.25 x 0.75 in. (Inside Slot 1.25 in.)
Description 2 Facet Edges
Sterility Non-sterile
Quantity 10/Pk.