missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Surgical Design Scalpel Blade Remover
Description
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Blade Remover |
| For Use With (Equipment) | Scalpels |
| Description | High Surface Finish |
| Disposable | Reusable |
| Autoclavable | Autoclavable |
| Color | Silver |