Learn More
Vector Laboratories STL, PL, Unconjugated, 5 mg AL-1013-2
Catalog No. 502045600
Encompass
Solanum Tuberosum (Potato) Lectin (STL, PL), Unconjugated, 5 mg, binds oligomers of N-acetylglucosamine and some bacterial cell wall oligosaccharides containing N-acetylglucosamine and N-acetylmuramic acid. Although the carbohydrate binding specificity is similar to that of wheat germ agglutinin and Datura stramonium lectin, several differences have been reported for potato lectin.
Catalog No. 502045600 Supplier Vector Laboratories Supplier No. AL10132
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Solanum Tuberosum (Potato) Lectin (STL, PL), Unconjugated, 5 mg, binds oligomers of N-acetylglucosamine and some bacterial cell wall oligosaccharides containing N-acetylglucosamine and N-acetylmuramic acid. Although the carbohydrate binding specificity is similar to that of wheat germ agglutinin and Datura stramonium lectin, several differences have been reported for potato lectin.