Learn More
Abcam Inc SB 203580 hydrochloride, p38 MAPK inhibitor, 10MG
Catalog No. 502707769
Encompass
MW 413.9 Da, Purity >98%. Water soluble, cell-permeable p38 MAPK inhibitor. Shows selectivity over many other kinases but also shown to have some inhibitory activity against GAK, CK1 , RIP2 c-Raf and GSK3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502707769 Supplier Abcam Inc Supplier No. AB12023510MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 413.9 Da, Purity >98%. Water soluble, cell-permeable p38 MAPK inhibitor. Shows selectivity over many other kinases but also shown to have some inhibitory activity against GAK, CK1 , RIP2 c-Raf and GSK3.
The product is subject to the following: Abcam Restricted Use Statement