Learn More
Abcam Inc SB 202190, cell-permeable p38 MAPK inhibitor, 10MG
Catalog No. 502707405
Encompass
MW 331.3 Da, Purity >99%. Highly selective, potent and cell-permeable p38 MAPK inhibitor (IC values are 50 and 100 nM at SAPK2a/p38 and SAPK2b/p382, respectively). Selective over a wide range of other kinases. Apoptosis and autophagy inducer. 3D Growth matrix component and additive for long term growth; also component of gastric organoid culture media.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502707405 Supplier Abcam Inc Supplier No. AB12063810MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 331.3 Da, Purity >99%. Highly selective, potent and cell-permeable p38 MAPK inhibitor (IC values are 50 and 100 nM at SAPK2a/p38 and SAPK2b/p382, respectively). Selective over a wide range of other kinases. Apoptosis and autophagy inducer. 3D Growth matrix component and additive for long term growth; also component of gastric organoid culture media.
The product is subject to the following: Abcam Restricted Use Statement