Learn More
Koehler™ Instrument Saybolt Viscometer Tube and Cleaner
Catalog No. 22280284
Change view
Click to view available options
Product Type:
Viscometer Tube
Catalog No. Product Type
22280284 Viscometer Tube
1 options
Catalog No. 22280284 Supplier Koehler™ Instrument Supplier No. K22009
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Order Info
Case of 1.
Specifications
Product Type Viscometer Tube
For Use With (Equipment) Saybolt Viscometer, Lubricants, Insulating Oils and Heater Fuel Grades
Certifications/Compliance ASTM D88, E102
Material Brass