Learn More
Imeb Inc Pathology Bone Band Saw Optional Saw Blade with Standard Teeth
Catalog No. NC0907435
Encompass
Saw Blade with Standard Teeth. Toothed Blade .1380" wide X .031" X 14tpi. IMEB Bone Band Saw sold separately. The IMEB, Inc. Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
Catalog No. NC0907435 Supplier Imeb Inc Supplier No. SF301
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Saw Blade with Standard Teeth. Toothed Blade .1380" wide X .031" X 14tpi. IMEB Bone Band Saw sold separately. The IMEB, Inc. Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.