missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Savory Oil, Summer Variety, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 25 mL |
| Grade | Reagent |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Savory Oil, Summer Variety |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.