missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
$86,979.78 - $114,075.31
| Catalog Number | Mfr. No. | Description | Electrical Requirements | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Electrical Requirements | Price | Quantity & Availability | ||||
|
13875337
|
Thermo Scientific™
SPD300DDA115 |
SpeedVac SPD300DDA Vacuum concentrator | 115 V 60 Hz |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
13875379
|
Thermo Scientific™
SPD300DDAA115 |
SpeedVac SPD300DDA Vacuum Concentrator Kit-No rotor | 115 V 60 Hz |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
13875375
|
Thermo Scientific™
SPD300P1115 |
SpeedVac SPD300DDA Vacuum Concentrator Kit | 115 V 60 Hz |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
13875377
|
Thermo Scientific™
SPD300P2115 |
SpeedVac SPD300DDA Vacuum Concentrator Kit | 115 V 60 Hz |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
- For strong acids and bases used in combinatorial chemistry