missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD120 Vacuum Concentrator and Kits
Description
These modular vacuum concentrators provide flexibility for your benchtop space. They are ideal for evaporation of non-aggressive organics. Choose from one of 2 kits available: SPD120P1 SpeedVac kit for standard evaporation or SPD120 P2 SpeedVac™ kit for lyophilization. All SpeedVac kits contain all the necessary components under one model number.
Space-saving design:
- Automatic bleeder valve for un-attended start/stop
- Digital display of parameters for easy, reproducible selection
- Three user defined programs that can be modified and saved
- Independent timers for both heat and run and audible alarm when run is completed
- PTFE-coated chamber for superior chemical resistance and easy cleaning
- Match up with an appropriate cold trap, vacuum pump or chemical trap for a complete concentration/drying system
Product Features:
- Basic molecular biology applications with increased microplate capacity
- Simple display and programming
- Six programs available
- Enhanced evaporation model, organic compatible
- Radiant glass cover to accelerate evaporation and reduce drying time
- Vacuum end point mode terminates run when preset vacuum level is reached
- Digital display of vacuum, temperature and time section
- Latched lid and lid lock indicator ensuring user safety during operation
SpeedVac SPD120 P1 Kit includes:
- SpeedVac concentrator
- Universal vacuum system (UVS450) including an oil-free vacuum pump and a -50 °C refrigerated vapor trap
- Glass condensation flask (GCF400)
- 40 x 1.5 mL rotor (RH40-11)
- Tubing package including 12’ of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
- CryoCool heat transfer fluid (1 L)
SpeedVac SPD120 P2 Kit includes:
- SpeedVac concentrator, basic molecular biology
- -105 ºC, ultra-low temperature refrigerated vapor trap
- VPL120 high vacuum pump
- 40 x 1.5 mL rotor (RH40-11)
- Universal tubing package (TFK100)
- Digital vacuum gauge
- Chemical trap kit for use with disposable cartridge
- Disposable cartridge with color indicator for trapping acids and water vapor
- Glass condensation flask
- Vacuum pump lubricating oil
- CryoCool™ heat transfer fluid (1 L)
Solvent and solvent combinations
- Acetonitrile
- Ethanol
- Isopropanol
- Methanol
- Water
- Acetonitrile and water
- Ethanol and water
- Methanol and water
- Ammonium hydroxide
Typical applications:
- Drying biomolecules in HPLC fractions
- Evaporating purification solvents
- Drying extracts of natural products in drug assays
- Lyophillization
Specifications
Specifications
| Includes | Kit for Standard Evaporation |
| Certifications | UL Listed to US and Canadian requirements. CE marked |
| Description | SpeedVac SPD120 Vacuum Concentrator Kit |
| Electrical Requirements | 115 V 60 Hz |