missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ SPD120 Vacuum Concentrator and Kits

Catalog No. 13875341
Change view
Click to view available options
Includes:
Kit for Standard Evaporation
Kit for Standard Evaporation Including Lyophilization
Vacuum Concentrator
Electrical Requirements:
115 V 60 Hz
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Electrical Requirements
13875341 Vacuum Concentrator 115 V 60 Hz
13875363 Kit for Standard Evaporation 115 V 60 Hz
13875365 Kit for Standard Evaporation Including Lyophilization 115 V 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13875341 Supplier Thermo Scientific™ Supplier No. SPD120115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SpeedVac™ SPD120 vacuum concentrator and kits.

These modular vacuum concentrators provide flexibility for your benchtop space. They are ideal for evaporation of non-aggressive organics. Choose from one of 2 kits available: SPD120P1 SpeedVac kit for standard evaporation or SPD120 P2 SpeedVac™ kit for lyophilization. All SpeedVac kits contain all the necessary components under one model number.

Space-saving design:

  • Automatic bleeder valve for un-attended start/stop
  • Digital display of parameters for easy, reproducible selection
  • Three user defined programs that can be modified and saved
  • Independent timers for both heat and run and audible alarm when run is completed
  • PTFE-coated chamber for superior chemical resistance and easy cleaning
  • Match up with an appropriate cold trap, vacuum pump or chemical trap for a complete concentration/drying system

Product Features:

  • Basic molecular biology applications with increased microplate capacity
  • Simple display and programming
  • Six programs available
  • Enhanced evaporation model, organic compatible
  • Radiant glass cover to accelerate evaporation and reduce drying time
  • Vacuum end point mode terminates run when preset vacuum level is reached
  • Digital display of vacuum, temperature and time section
  • Latched lid and lid lock indicator ensuring user safety during operation

SpeedVac SPD120 P1 Kit includes:

  • SpeedVac concentrator
  • Universal vacuum system (UVS450) including an oil-free vacuum pump and a -50 °C refrigerated vapor trap
  • Glass condensation flask (GCF400)
  • 40 x 1.5 mL rotor (RH40-11)
  • Tubing package including 12’ of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
  • CryoCool heat transfer fluid (1 L)

SpeedVac SPD120 P2 Kit includes:

  • SpeedVac concentrator, basic molecular biology
  • -105 ºC, ultra-low temperature refrigerated vapor trap
  • VPL120 high vacuum pump
  • 40 x 1.5 mL rotor (RH40-11)
  • Universal tubing package (TFK100)
  • Digital vacuum gauge
  • Chemical trap kit for use with disposable cartridge
  • Disposable cartridge with color indicator for trapping acids and water vapor
  • Glass condensation flask
  • Vacuum pump lubricating oil
  • CryoCool™ heat transfer fluid (1 L)

Solvent and solvent combinations

  • Acetonitrile
  • Ethanol
  • Isopropanol
  • Methanol
  • Water
  • Acetonitrile and water
  • Ethanol and water
  • Methanol and water
  • Ammonium hydroxide

Typical applications:

  •  Drying biomolecules in HPLC fractions
  • Evaporating purification solvents
  • Drying extracts of natural products in drug assays
  • Lyophillization

Specifications

Model Medium capacity glass lid
Includes Vacuum Concentrator
Certifications UL Listed to US and Canadian requirements. CE marked
Description SpeedVac SPD120 Vacuum Concentrator
Electrical Requirements 115 V 60 Hz