Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Catalog No. 36792169
Change view
Click to view available options
Holds:
10 Centrifuge Tubes
100 (8 x 29 mm) Tubes
12 Scintillation Vials
120 Microtubes
24 Vials
32 Tubes
4 x 100 mL Pear-shaped Flasks
40 Microtubes
40 Tubes
6 x 50 mL Conical Tubes
60 Vials
64 Microtubes
72 Tubes
8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
36792169 24 Vials
20548107 100 (8 x 29 mm) Tubes
367815 10 Centrifuge Tubes
20548102 120 Microtubes
360737 12 Scintillation Vials
20548124 24 Vials
367819 32 Tubes
360727 40 Microtubes
360733 40 Tubes
20548121 4 x 100 mL Pear-shaped Flasks
360731 60 Vials
360738 64 Microtubes
367842 6 x 50 mL Conical Tubes
367802 72 Tubes
360735 8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 36792169 Supplier Thermo Scientific™ Supplier No. RH24 18
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
Specifications
Description 24-place; 18 x 52mm mini scint vials
Type Rotor
For Use With (Equipment) SPD1030, SPD120, SPD130, SPD140DDA
Holds 24 Vials
Model RH24-18
Rotor System Traditional