Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
Supplier:  Thermo Scientific™ RH1015
 View more versions of this product
Catalog No. 367815
Only null left
Description
Description
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
Specifications
Chemical Identifiers
Rotor
Traditional
Specifications
Centrifuge Tubes: 10 x 15mL conical, Medium capacity models
SPD1030, SPD120, SPD130, SPD140DDA
RH10-15
Rotor
10 Centrifuge Tubes
Traditional
Product Suggestions
Product Suggestions