Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
Supplier:  Thermo Scientific™ RH64 11
 View more versions of this product
Catalog No. 360738
Only null left
Description
Description
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
Specifications
Chemical Identifiers
Rotor
Traditional
Specifications
Microtubes: 64 x 1.5mL, Medium capacity models
SPD1030, SPD120, SPD130, SPD140DDA
RH64-11
1.5 mL
Rotor
64 Microtubes
Traditional
Product Suggestions
Product Suggestions