Learn More
Thermo Scientific™ Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Catalog No. p-7115396
Change view
Click to view available options
Electrical Requirements:
115 V 60 Hz
Catalog No. Electrical Requirements
13875349 115 V 60 Hz
1 options
Catalog No. 13875349 Supplier Thermo Scientific™ Supplier No. RVT450115
Only null left
Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT).   Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump. 
  • Rapid cooling time minimizes waiting period
  • Easy to remove glass flask for quick solvent changes
  • Rapidly cools to operating temperature
  • Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
  • Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
Specifications
Certifications/Compliance UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Description Refrigerated Vapor Cold Trap RVT450
Type Refrigerated Vapor Trap
Electrical Requirements 115 V 60 Hz