Learn More
Thermo Scientific™ Savant™ SpeedVac™ Medium Capacity Vacuum Concentrators for Combinatorial Chemistry Applications
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
SpeedVac SPD140DDA is a medium capacity vacuum concentrator used with a wide variety of solvents and offer resistance to TFA, DSMO and other aggressive solvents used in combinatorial chemistry applications. It is available in kit for ease of ordering.
SPD140DDA Features:
- All the product features of SPD130DLX PLUS:
- Radiant lamps within the chamber
- Heated lid for processing high boiling points (DMSO,DMF)
- Independent timers for both heat and run
- Audible alarm to alert user when operation is finished
- Built-in vacuum gauge and vacuum display
- Wider vapor port and tubing for enhanced vapor throughput - reducing drying times
- Adjustable vacuum level for preventing bumping of difficult solvent mixtures and optimizing drying times
- -5°C Cold Trap specially designed for trapping high boiling point solvents
Solvent and Solvent Combinations for SPD140P1 KIT
- Acetic acid
- Ammonium hydroxide
- Formic acid
- Hydrochloric acid
- TFA
- Methylene chloride and TFA
- Acetone and HCL
- Methylene chloride and MEOH
- DMSO and methanol
Solvent Combinations for SPD140 P2 KIT
- DMSO
- DMF
- Toluene
SPD140 P1 includes:
- Acid-Resistant SpeedVac™ Vacuum Concentrator
- -104°C Ultra-Low Temperature Refrigerated Vapor Trap
- Oil-Free ETFE/PFA 4-Headed Pump
- 40 x 1.5-2.0ml Glass Tubes Rotor (RH40-11)
- Glass Condensation Flask
- Tubing and Fitting Kit
- CryoCool™ Heat Transfer Fluid (1L)
SPD140P2 includes:
- Acid-Resistant SpeedVac™ Vacuum Concentrator
- -50°C Universal Vacuum Source
- 24 x 1 dram vials (15 x 45 mm) 4ml - dram vial Rotor (RH24-15)
- Chemical Trap
- Tubing and Fitting Kit
- Disposable Cartridge
- Glas Condensation Flask
- Vacuum Pump Lubricating Oil
- CryoCool™ Heat Transfer Fluid (1L)
SPD140 P1 and P2 kits are recommended for:
- Solid phase extractions
- Drying lipid extractions
- Flash chromatography fractions
- Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
- Compound management
- Evaporating dissolution solventsRotors are available for a wide variety of applications and sample formats. Please check the rotor list.
Typical applications SPD140DDA:
Requires:
- Drying lipid extraction
- Flash chromatography fractions
- Combinatorial Chemistry
- Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
- Removing ethyl acetate from samples for CAT assay
- Drug metabolites in solid phase extracts
- Analyte elution solvents for evaporation
Specifications
Specifications
| Type | Medium Capacity DDA |
| Certifications | UL Listed to US and Canadian requirements. CE marked |
| Description | SpeedVac SPD140DDA Vacuum Concentrator |
| Electrical Requirements | 115 V 60 Hz |