missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits with Installation

Get all the necessary components for HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications with these kits.

Supplier:  Thermo Scientific™ SPD2030A220INS

 View more versions of this product

Catalog No. 13875361INS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant™ SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac™ SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac™ SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).

Easy to Use:

  • Plug and play installation: Includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
  • Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
  • Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250 mL flasks
  • Audible alarm to alert user when operation is finished

Robust, Low-Maintenance Design:

  • Oil-free vacuum pump is virtually maintenance free
  • PTFE coated chamber protects the chamber from corrosion, while making cleaning easy

Enhanced Evaporation:

  • Faster sample drying provided with radiant cover
  • Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
  • Reduced length vapor path from concentrator to refrigerated vapor trap accelerates evaporation
  • Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates

Solvent and Solvent Combinations:

  • Acetonitrile
  • Ethanol
  • Methanol
  • Methylene Chloride
  • Water
  • Other organic solvents

SPD1030A and SPD2030A includes:

  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD1030P1 Kit includes:

  • Fully Integrated SpeedVac system
  • 40 × 1.5 mL rotor (RH40-11)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD2030P1 kit

  • Integrated SpeedVac system
  • 200 × 1.5 mL rotor (RH200-12)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)
Specifications

Specifications

<10 Torr (13 mbar, 1.3k Pa)
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
113 kg
SPD2030
Ambient to 80°C
SpeedVac SPD2030 Integrated Vacuum Concentrator (rotor- order separately)
8 A
31 x 28 x 19 in. (79 x 71 x42 cm)
250 lb.
Diaphragm pump, cold trap and vacuum gauge
Vacuum Concentrator
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
PTFE-coated Aluminum
220 V 60 Hz
94 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions