Learn More
Thermo Scientific™ Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits with Installation
Get all the necessary components for HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications with these kits.
Supplier: Thermo Scientific™ SPD1030A115INS
Description
Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant™ SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac™ SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac™ SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).
Easy to Use:
- Plug and play installation: Includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
- Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
- Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250 mL flasks
- Audible alarm to alert user when operation is finished
Robust, Low-Maintenance Design:
- Oil-free vacuum pump is virtually maintenance free
- PTFE coated chamber protects the chamber from corrosion, while making cleaning easy
Enhanced Evaporation:
- Faster sample drying provided with radiant cover
- Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
- Reduced length vapor path from concentrator to refrigerated vapor trap accelerates evaporation
- Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates
Solvent and Solvent Combinations:
- Acetonitrile
- Ethanol
- Methanol
- Methylene Chloride
- Water
- Other organic solvents
SPD1030A and SPD2030A includes:
- Integrated SpeedVac system
- Glass condensation Flask (GCF400)
SPD1030P1 Kit includes:
- Fully Integrated SpeedVac system
- 40 × 1.5 mL rotor (RH40-11)
- CryoCool™ 1 L heat transfer fluid
- Glass condensation Flask (GCF400)
SPD2030P1 kit
- Integrated SpeedVac system
- 200 × 1.5 mL rotor (RH200-12)
- CryoCool™ 1 L heat transfer fluid
- Glass condensation Flask (GCF400)
Chemical Identifiers
| UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives. | |
| Ambient to 80°C | |
| 8 A |
| <10 Torr (13 mbar, 1.3k Pa) | |
| PTFE-coated Aluminum |
Specifications
| SPD1030 | |
| 198 lb. | |
| Diaphragm pump, cold trap and vacuum gauge | |
| Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates. | |
| Ambient to 80°C | |
| SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor- order separately) | |
| 8 A | |
| 69 kg |
| Vacuum Concentrator | |
| UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives. | |
| 90 kg | |
| <10 Torr (13 mbar, 1.3k Pa) | |
| PTFE-coated Aluminum | |
| 115 V 60 Hz | |
| 139 lb. | |
| 25 x 26 x 16 in. (64 x 66 x 41 cm) |