missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Integrated Vacuum Concentrator Systems and Kits

Catalog No. 13875357
Change view
Click to view available options
Model:
SPD1030
SPD2030
Includes:
Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor
Diaphragm Pump, Cold Trap, Vacuum Gauge
Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor
Vacuum Concentrator
Electrical Requirements:
115 V 60 Hz
220 V 60 Hz
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes Electrical Requirements
13875357 SPD2030 Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor 220 V 60 Hz
13875339 SPD1030 Vacuum Concentrator 115 V 60 Hz
13875359 SPD1030 Diaphragm Pump, Cold Trap, Vacuum Gauge 115 V 60 Hz
13875355 SPD1030 Diaphragm Pump, Cold Trap, Vacuum Gauge, RH40-11 Rotor 115 V 60 Hz
13875361 SPD2030 Diaphragm Pump, Cold Trap, Vacuum Gauge 220 V 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13875357 Supplier Thermo Scientific™ Supplier No. SPD2030P1220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Get all the necessary components for HPLC fractions with water/0.1% trifluoroacetic acid and reversed-phase SPE eluants applications with these kits.

Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).

Easy to Use:

  • Plug and play installation:. Includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
  • Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
  • Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250-ml flasks
  • Audible alarm to alert user when operation is finished

Robust, Low-Maintenance Design:

  • Oil-free vacuum pump is virtually maintenance free
  • PTFE coated chamber protects the chamber from corrosion, while making cleaning easy

Enhanced Evaporation:

  • Faster sample drying provided with radiant cover
  • Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
  • Reduced length vapor path from concentrator to refrigerated vapor trap accelerates evaporation
  • Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates

Solvent and Solvent Combinations:

  • Acetonitrile
  • Ethanol
  • Methanol
  • Methylene Chloride
  • Water
  • Other organic solvents

SPD1030A and SPD2030A includes:

  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD1030P1 Kit includes:

  • Fully Integrated SpeedVac system
  • 40 × 1.5mL rotor (RH40-11)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)

SPD2030P1 kit

  • Integrated SpeedVac system
  • 200 × 1.5mL rotor (RH200-12)
  • CryoCool™ 1L heat transfer fluid
  • Glass condensation Flask (GCF400)

Specifications

Model SPD2030
Includes Diaphragm Pump, -50°C Cold Trap, Vacuum Gauge, RH40-11 Rotor
Shipping Weight (Metric) 136 kg
Certifications UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Max. Vacuum <10 Torr (13 mbar, 1.3k Pa)
Holds Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Shipping Weight (English) 260 lb.
Temperature Range (Metric) Ambient to 80°C
Chamber Material PTFE-coated Aluminum
Description SPD2030 Integrated System Kit
Electrical Requirements 220 V 60 Hz
Amperage 8 A
Weight (Metric) 98 kg
Dimensions (L x W x H) 31 x 28 x 19 in. (79 x 71 x42 cm)
Show More Show Less