missing translation for 'onlineSavingsMsg'
Learn More

Contec™ SATWIPES™ Wipes

Contec™ SATWIPES™Wipes consist of nonwoven presaturated wipes dispensed from a center-pull roll in a reusable canister.

Supplier:  Contec™ SW420045

Catalog No. 296367898


Only null left
Explore available promotions

Description

Description

  • Solvent-specific color-coded labeling for easy solvent identification
  • Wipes are dispensed from center-pull perforated rolls with self-closing lids for added convenience
Specifications

Specifications

Pre-saturated Wipe
15 cm
23 cm
Not Autoclavable
15 x 23 cm (6 x 9 in.)
Non-sterile
6 in.
9 in.
Non-woven Cellulose, Lyocell
Single-use
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions