Learn More
Bioscience International SAS Super 180 Bioaerosol Sampler, High-Volume Air Sampling, Autoclavable Stainless Steel, 1.7 kg
Catalog No. NC9381239
Encompass
The SAS Super 180 Bioaerosol Sampler is a high-volume microbial air sampler designed for precise assessment of air quality in scientific and industrial settings. It collects airborne microorganisms efficiently, complying with major standards. Key features include an airflow rate of 180 L/min, programmable sampling options, and a durable, autoclavable construction using AISI 316 stainless steel, ensuring reliable performance and easy maintenance. This makes it ideal for applications like environmental monitoring and cleanroom validation.
Catalog No. NC9381239 Supplier Bioscience International Supplier No. 19121
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The SAS Super 180 Bioaerosol Sampler is a high-volume microbial air sampler designed for precise assessment of air quality in scientific and industrial settings. It collects airborne microorganisms efficiently, complying with major standards. Key features include an airflow rate of 180 L/min, programmable sampling options, and a durable, autoclavable construction using AISI 316 stainless steel, ensuring reliable performance and easy maintenance. This makes it ideal for applications like environmental monitoring and cleanroom validation.