Learn More
Sartorius Water-Jet Pump
Catalog No. 14555894
Change view
Click to view available options
Flow Rate:
Variable
Catalog No. Flow Rate
14555894 Variable
1 options
Catalog No. 14555894 Supplier Sartorius Supplier No. 16611
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Water-Jet Pump is a maintenance-friendly and performant pump for smooth and reliable filtration.
  • With G 3/4 female thread
Specifications
Max. Vacuum Variable
Includes G 3/4 female thread
Flow Rate Variable
Pump Type Vacuum System
Applications Sample Filtration in Microbiology
For Use With (Application) Connection to Water tap with G 3/4 male thread