Learn More
Sartorius Sterisart™ NF Sterility Testing System
Provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices. Sartorius Sterisart™ NF Sterility Testing System is optimal for use with isolators.
$428.81 - $1,089.59
Specifications
Capacity (English) 4 oz.
Capacity (Metric) 120 mL
For Use With (Application) Sterility Testing
Model Sterisart Consumable
Temperature (English) 122°F
View More Specs
Products 14
Catalog Number Mfr. No. Sterilization Method Sample Composition Includes Sampling Option Price Quantity & Availability  
Catalog Number Mfr. No. Sterilization Method Sample Composition Includes Sampling Option Price Quantity & Availability  
14555830
View Documents Promotion Details
Sartorius
16466 GSD
Gamma Irradiation Liquids Long dual-needle spike, sterile vented, septum Septum
Pack of 10 Each for $1,089.59
Only null left
14555829
View Documents Promotion Details
Sartorius
16466 GBD
Gamma Irradiation Liquids Long dual-needle spike, sterile vented No Septum
Pack of 10 Each for $1,002.42
Only null left
14555833
View Documents Promotion Details
Sartorius
16467 GBD
Gamma Irradiation Liquids Long needle, by-packed sterile venting needle No Septum
Pack of 10 Each for $1,002.42
Only null left
14555834
View Documents Promotion Details
Sartorius
16468 GBD
Gamma Irradiation Liquids Luer (Lock) connection, by-packed long needle, sterile venting No Septum
Pack of 10 Each for $1,002.42
Only null left
14555841
View Documents Promotion Details
Sartorius
16478 GBD
Gamma Irradiation Liquids Female luer lock connector No Septum
Pack of 10 Each for $1,002.42
Only null left
14555835
View Documents Promotion Details
Sartorius
16469 GBD
Gamma Irradiation Liquids Universal Luer adapter, long dual-needle spike, sterile vented No Septum
Pack of 10 Each for $1,002.42
Only null left
14555836
View Documents Promotion Details
Sartorius
16470 GBD
Gamma Irradiation Liquids Tubing system w/ two dual-needle metal spikes of different length No Septum
Pack of 10 Each for $428.81
Only null left
14555840
View Documents Promotion Details
Sartorius
16477 GBD
Gamma Irradiation Liquids Long needle, side opening w/solid pointed tip No Septum
Pack of 10 Each for $1,002.42
Only null left
14555838
View Documents Promotion Details
Sartorius
16475 GBD
Gamma Irradiation Easily Dissolved Powders Two dual-needle spikes of different length, sterile vented No Septum
Pack of 10 Each for $1,015.08
Only null left
14555839
View Documents Promotion Details
Sartorius
16476 GBD
Gamma Irradiation Liquids Short dual-needle spike, sterile vented No Septum
Pack of 10 Each for $1,002.42
Only null left
14556140
View Documents Promotion Details
Sartorius
16467 GSD
Gamma Irradiation Liquids Long needle, sterile venting needle, septum Septum
Pack of 10 Each for $1,089.59
Only null left
14556145
View Documents Promotion Details
Sartorius
16476 GSD
Gamma Irradiation Liquids Short dual-needle spike, sterile vented, with septum Septum
Pack of 10 Each for $1,089.59
Only null left
14556143
View Documents Promotion Details
Sartorius
16469 GSD
Gamma Irradiation Liquids Universal Luer adapter, long dual-needle spike, sterile vented, septum Septum
Pack of 10 Each for $1,089.59
Only null left
14556144
View Documents Promotion Details
Sartorius
16475 GSD
Gamma Irradiation Easily Dissolved Powders Two dual-needle spikes of different length, one sterile vented w/septum Septum
Pack of 10 Each for $1,089.59
Only null left
Description
Description
International pharmacopoeias require the complete sterility of pharmaceutical products that are injected into the blood stream or that otherwise enter the body below the skin surface. As a manufacturer of such products, you are required to supply proof of sterility of the final product batch. Sterisart™ NF is a completely closed system for the sterility testing of pharmaceutical products. It is based on the membrane filter method, however it eliminates the procedure of manipulating the filters. By this the main risk of a secondary contamination and false positive results is eliminated. A peristaltic pump transfers the sample into the filtration units, and after rinsing, the filtration units are filled with media and used for incubation of the filters without any contact to the environment.
  • reliable Sartochem™ membrane
  • high retention of microbes
  • low adsorption
  • high mechanical stability
  • pre-installed color-coded tube clamps
  • easy-to-read graduated marks
  • user-friendly (several practical adapters available)
  • product-/lot number identification
  • gas-impermeable packaging for protection against sterilants
Specifications
Specifications
4 oz.
Sterility Testing
122°F
15.7 cm
3 bar at 20°C
120 mL
Sterisart Consumable
50°C
0.45 μm
Sterile