Learn More
Sartorius Sterility Testing Reusable System: Ampoule Breaker
Description
- With two blocks for large or small ampoules
- Simply insert ampoule in breaker and snap
Specifications
Specifications
| Disposable | Reusable |
| For Use With (Equipment) | Reusable sterility test system |
| Sterility | Non-sterile |
| Type | Ampoule Breaker |