Learn More
Sartorius Pressure Heads for Vivaspin™ 20 and Vivacell™ 70, 100, 250
Catalog No. 14558299
Change view
Click to view available options
For Use With:
Vivacell™ 100
Vivaspin™ 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
14558299 Vivaspin™ 20
14558303 Vivacell™ 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14558299 Supplier Sartorius Supplier No. VCA200
For various Vivacell models
The Vivaspin™ 20 is an ultrafiltration unit with unique twin membrane for sample concentration with high process flexibility: centrifugation or centrifugation of pressurized units, as well as pressure filtration in stationary benchtop mode.
Specifications
For Use With Vivaspin™ 20
Type Pressure Head