missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Nutrient Pad Sets (NPS) 47mm diameter

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

$303.12 - $311.37

Specifications

Form Dehydrated
Manufacturing Origin Germany
Sterility Sterile
Test Type Microbial Detection
Packaging Type Individually Packaged
View More Specs

Products 25
Catalog Number Mfr. No. Color Grid Color Pore Size Media Type Price Quantity  
Catalog Number Mfr. No. Color Grid Color Pore Size Media Type Price Quantity  
14-555-669
View Documents Promotion Details
Sartorius
1405147N
Green Dark Green 0.45μm Azide N/A
Only null left
 
14-555-670
View Documents Promotion Details
Sartorius
1405347N
White Green 0.45μm Endo N/A
Only null left
 
14-555-674
View Documents Promotion Details
Sartorius
1405547N
Green Dark Green 0.45μm Standard TTC N/A
Only null left
 
14-555-678
View Documents Promotion Details
Sartorius
1405647N
White Green 0.45μm Tergitol TTC N/A
Only null left
 
14-555-679
View Documents Promotion Details
Sartorius
1405747N
Green Dark Green 0.45μm Bismuth Sulfite N/A
Only null left
 
14-555-681
View Documents Promotion Details
Sartorius
1405847N
Gray White 0.65μm Bismuth Sulfite N/A
Only null left
 
14-555-684
View Documents Promotion Details
Sartorius
1405947N
White Green 0.45μm VLB-S7-S N/A
Only null left
 
14-555-688
View Documents Promotion Details
Sartorius
1406247N
Green Dark Green 0.45μm Orange Serum N/A
Only null left
 
14-555-690
View Documents Promotion Details
Sartorius
1406347N
Green Dark Green 0.45μm Caso N/A
Only null left
 
14-555-691
View Documents Promotion Details
Sartorius
1406447N
Green Dark Green 0.45μm Standard N/A
Only null left
 
14-555-696
View Documents Promotion Details
Sartorius
1406647N
White Green 0.45μm Glucose Tryptone N/A
Only null left
 
14-555-700
View Documents Promotion Details
Sartorius
1406847N
White Green 0.45μm m-FC N/A
Only null left
 
14-555-703
View Documents Promotion Details
Sartorius
1406947N
Gray White 0.65μm Sabouraud Agar N/A
Only null left
 
14-555-709
View Documents Promotion Details
Sartorius
1407547N
White Green 0.45μm Cetrimide N/A
Only null left
 
14-555-708
View Documents Promotion Details
Sartorius
1407447N
White Green 0.45μm Chapman N/A
Only null left
 
14-555-711
View Documents Promotion Details
Sartorius
1407647N
Green Dark Green 0.45μm TGE N/A
Only null left
 
14-555-713
View Documents Promotion Details
Sartorius
1407947N
Green Dark Green 0.45μm Jus de Tomate N/A
Only null left
 
14-555-721
View Documents Promotion Details
Sartorius
1408447N
Green Dark Green 0.45μm R2A N/A
Only null left
 
14-555-723
View Documents Promotion Details
Sartorius
1408647N
Gray White 0.8μm Malt Extract N/A
Only null left
 
14-555-724
View Documents Promotion Details
Sartorius
1408647CCN
White Black 0.45μm Malt Extract N/A
Only null left
 
14-555-728
View Documents Promotion Details
Sartorius
1409647N
White Green 0.45μm Acid-tolerant N/A
Only null left
 
14-555-726
View Documents Promotion Details
Sartorius
1408947N
Green Dark Green 0.45μm Wallerstein N/A
Only null left
 
14-555-725
View Documents Promotion Details
Sartorius
1408747N
Gray White 0.45μm Chromocult N/A
Only null left
 
14-555-727
View Documents Promotion Details
Sartorius
1409547N
Gray White 0.8μm Standard N/A
Only null left
 
14-555-729
View Documents Promotion Details
Sartorius
1409747N
Gray White 0.45μm MacConkey N/A
Only null left
 
Description

Description

Sartorius Nutrient Pad Sets are used in the membrane filter methods. The membrane filter method allows large sample volumes with low microbial count to be tested. Inhibitors, such as essential oils or disinfectants, can be rinsed out after filtration, before incubation.Practical and easy to handle, Nutrient Pads reduce labor and simplify many microbiological testing procedures. Microorganisms require the appropriate culture media for optimal growth.Nutrient Pad Sets are sterile, dehydrated culture media. Once they are moistened with 3.0 - 3.5ml of sterile and demineralized (or destilled) water they are ready-to-use immediately. The level of moisture is optimal when an excess ring of water surrounding the NPS is visible.All Nutrient Pad Set types are supplied with the appropriate membrane filters, which are also individually, sterile packaged. The membrane filters tailored to meet the special requirements of microbial detection are available with 47 or 50mm diameter. The standard NPS box contains 100 sterile Nutrient Pads, each of which is individually inserted in a petri dish and sterilized. Ten each of these petri dishes are sealed in an aluminum bag. This special packaging in bags protects the sensitive formula constituents of the Nutrient Pad during transport and storage from fluctuations in humidity and temperature. As a result, it guarantees the high quality of NPS throughout their entire shelf life up to 24 months. More than 30 different Nutrient Pad Set types are available to meet the diverse objectives of microbiological analysis. Beyond the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.

  • trouble-free storage up to 24 month
  • economical by time- and cost-saving
  • according to international standards and references
  • easy handling
  • validation guide available
  • consistent quality
  • highly versatile
Specifications

Specifications

Dehydrated
Sterile
Individually Packaged
4.7 cm
Germany
Microbial Detection
1.85 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'