Learn More
Sartorius Glass Fiber Filters
Description
- Soft texture with high suction power/Nonhydrophilic/Binder-free
Specifications
Specifications
| For Use With (Equipment) | Moisture Analyzer |
| Material | Glass Fiber |
| Type | Filter |