Learn More
Sartorius Funnels for Combisart™
Catalog No. 14555951
Change view
Click to view available options
Product Type:
Funnel
Funnel Lid
Includes:
Locking Clamp
Small Central Air Port
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
14555951 Funnel Lid Small Central Air Port
14555948 Funnel Locking Clamp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14555951 Supplier Sartorius Supplier No. 6981063
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reusable stainless steel Combisart™ Funnels for microbial enumeration and particle counting.
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
Specifications
Capacity (Metric) 100 mL
Material Stainless Steel
For Use With (Equipment) Combisart Funnel
Includes Small Central Air Port
Quantity 1 Ea.
Product Type Funnel Lid