Learn More
Sartorius Funnel
Catalog No. 14555953
Change view
Click to view available options
For Use With (Equipment):
Combisart System
Catalog No. For Use With (Equipment)
14555953 Combisart System
1 options
Catalog No. 14555953 Supplier Sartorius Supplier No. 6981065
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reusable stainless steel funnels for microbial enumeration and particle counting.
  • Specially designed for use in microbiological quality assurance, these three filter holders differ only in their capacity
  • The reusable funnels feature a special locking clamp that simplifies the positioning and removal of the membrane filter, ensuring secure lock-down
  • For traceability, each filter funnel has an individual serial number
Specifications
Autoclavable Autoclavable
Capacity (Metric) 100 mL
For Use With (Equipment) Combisart System
Type Funnel
For Use With (Application) Colony or particle counting
Includes Closure clamp
Material Stainless Steel
Diameter (Metric) Filter 4.7/5 cm