missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Disposable Stands for Concentrators

Catalog No. 14558391
Change view
Click to view available options
Positions:
4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
14558391 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14558391 Supplier Sartorius Supplier No. VPA002

3 - 20mL Vivapore™ are ready-to-use static concentrators for fast and affordable concentration of samples. Extremely simple and user-friendly. Stand and pipettes included in packs with 4 and 30 devices. Sample concentration without additional equipment.

Vivapore™ 5 and Vivapore™ 10/20 are a ready-to-use, disposable ultrafiltration devices for samples up to 20 ml initial volume. They are optimally utilized for concentrating macromolecules in clinical specimens such as urine, cerebrospinal fluid (CSF) or other biological solutions. Two different devices are available: Vivapore™ 5 for concentration of 5 ml samples and Vivapore™ 10/20 for concentration of 10 ml samples expandable with an expansion reservoir to 20 ml. With no need for additional equipment, solvent absorption is the most economic and user friendly concentration technique available for clinical applications. Just fill the unit with the solution to be concentrated, wait for the desired concentration level to be achieved and then pipette the concentrated sample from the dead-stop reservoir at the bottom of the concentrator. Vivapore™ static concentrators are offered as separate chambers together with disposable plastic stands for up to four units. Contrary to other, larger products with multiple, adjacent concentration channels, there is no risk of cross-contamination between individual Vivapore™ units. Vivapore™ 5 and 10/20 are frequently used for urine concentration prior to subsequent analysis of Bence-Jones proteins by gel or capillary electrophoresis.

  • Optimal for clinical specimen concentration e.g. urine High-flow 7.5 kDa PES membrane
  • Separate chambers / no cross contamination
  • For up to 20 ml sample volume

Specifications

Autoclavable Not Autoclavable
Color White/White
Holds Solvent Absorption Concentrators
Material Stainless Steel
Positions 4
Shape Horizontal
Stackable Yes
Sterility Non-sterile
Product Type Disposable Stand
Shelves 4 Tiers
Height (Metric) 6.4 cm
Width (Metric) 7 cm
Show More Show Less