Learn More
Sartorius Sartobind™ Lab LC System Adapter Kit
Description
- High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes
- Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity
- Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration
- Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system
- Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production
Specifications
Specifications
| Type | Adapter |
| Material | PEEK |