Learn More
ACROBiosystems Inc SARS-CoV-2 S2 protein, His Tag (BA.1/Omicron) (S2N-C52Hf) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (N764K, D796Y, N856K, Q954H, N969K, L981F, F817P, A892P, A899P, A942P, K986P, V9
Catalog No. 502253601
Encompass
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 502253601 Supplier ACROBiosystems Inc Supplier No. S2NC52HF100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.